Learn More
Biomatik Corporation Mouse Granulocyte Chemotactic Protein 2 (GCP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09672
Encompass_Preferred
Mouse Granulocyte Chemotactic Protein 2 (GCP2) CLIA Kit for detecting GCP2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.07pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q921G8. Gene ID: 74237. Target Synonyms: CXCL6; SCYB6; GCP2; CKA-3; Chemokine (C-X-C-Motif) ligand 6; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 6; Chemokine alpha 3; Small-inducible cytokine B6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8470