Learn More
American Research Products Inc Mouse Granulocyte Colony Stimu
Catalog No. 5000328945
Encompass
Mouse Granulocyte Colony Stimulating Factor G-CSF ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28945 Supplier American Research Products Inc Supplier No. CSBE04564M
Mouse Granulocyte Colony Stimulating Factor G-CSF ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich