Learn More
American Research Products Inc Mouse Granulocyte Macrophage C
Catalog No. 5000328765
Encompass
Mouse Granulocyte-Macrophage Colony Stimulating Factor GM-CSF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28765 Supplier American Research Products Inc Supplier No. CSBE04569M
Mouse Granulocyte-Macrophage Colony Stimulating Factor GM-CSF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich