Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse GM-CSF Recombinant Protein

Catalog No. PIRMGMCSF20
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRMGMCSF20 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRMGMCSF20 Supplier Invitrogen™ Supplier No. RMGMCSF20
Add to Cart
Add to Cart

Recombinant Protein

This product cross-reacts with rat, mouse, trout, bacteria, leech, and human GM-CSF. Use: Mouse GM-CSF exerts activity in the range of 0.05 to 0.5 ng/mL for most in vitro applications. Reconstitution and Dilution: Reconstitute in distilled water to a concentration of 100 μg/mL. After reconstitution, dilute cytokine in a neutral buffer containing carrier protein such as 5-10% FBS or 1-4% high purity BSA (fatty acid free fraction V). Lyophilized product is stable until expiration if stored desiccated below 0°C. Cytokines diluted as indicated above should be divided into aliquots and stored at -70°C. Recombinant proteins should be stored at concentrations >500 ng/mL to reduce loss of activity.

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01587
For Use With (Application) Control, ELISA, Immunoprecipitation, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 12981
Molecular Weight (g/mol) 14.2 kDa
Name Mouse GM-CSF
Quantity 20 μg
Endotoxin Concentration <1 EU/ μg
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Common Name GM-CSF
Gene Symbol CSF2
Biological Activity ED50 ≤ 0.05 ng/mL; determined by the dose-dependent proliferation of mouse FDC-P1 cells.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Purity or Quality Grade >98% by SDS-PAGE and RP-HPLC
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.