Learn More
Sino Biological Mouse GREM1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50016R117100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse GREM1 (Catalog#50016-M08B; O70326; Met1-Asp184).
Catalog No. 50-237-2782