Learn More
Sino Biological Anti-GRM1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106527T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse GRM1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1038