Learn More
Sino Biological Mouse GRN MAb 20ul (Sino Biological)
Supplier:  Sino Biological GMP11725HNAE20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Progranulin / Granulin (rM Progranulin / Granulin; Catalog#50396-M08H; NP_032201.2; Met1-Leu589).
Catalog No. 50-237-4556