Learn More
Biomatik Corporation Mouse Growth Arrest And DNA Damage Inducible Protein Gamma (GADD45g) ELISA Kit
Supplier:  Biomatik Corporation EKU04576
Encompass_Preferred
Mouse Growth Arrest And DNA Damage Inducible Protein Gamma (GADD45g) ELISA Kit for detecting GADD45g in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z111. Target Synonyms: CR6; DDIT2; GADD45gamma; GRP17; Gadd-Related Protein,17 kD; Cytokine-responsive protein CR6; DNA damage-inducible transcript 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8945