Learn More
American Research Products Inc Mouse growth arrest specific g
Catalog No. 5000329573
Encompass
Mouse growth arrest-specific gene-6 (gas-6) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 17 086 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29573 Supplier American Research Products Inc Supplier No. CSBE14145M
Mouse growth arrest-specific gene-6 (gas-6) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 17 086 pg/mL Sample serum plasma tissue homogenates Method Sandwich