Learn More
Biomatik Corporation Mouse Growth Differentiation Factor 11 (GDF11) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10283
Encompass_Preferred
Mouse Growth Differentiation Factor 11 (GDF11) High-sensitive ELISA Kit for detecting GDF11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.21pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1W4. Gene ID: 14561. Target Synonyms: BMP11; Bone morphogenetic protein 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8353