Learn More
American Research Products Inc Mouse Growth differentiation f
Catalog No. 5000327134
Encompass
Mouse Growth/differentiation factor 11 (GDF11) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27134 Supplier American Research Products Inc Supplier No. CSBEL009344MO
Mouse Growth/differentiation factor 11 (GDF11) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich