Learn More
Raybiotech Inc C-Series Mouse Growth Factor Antibody Array 2 Kit.
Supplier:  Raybiotech Inc AAMGF24
Encompass_Preferred
Detects 20 Mouse Growth Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Amphiregulin, bFGF, EGF, GCSF, GM-CSF, HGF, IGF-1, IGF-2, IGFBP-2, IGFBP-3, IGFBP-5, IGFBP-6, IL-2, IL-7, M-CSF, SCF, VEGF-A, VEGF-D, VEGFR1, VEGFR3
Catalog No. 50-194-8732