Learn More
Biomatik Corporation Mouse Growth Hormone (GH) CLIA Kit
Supplier:  Biomatik Corporation EKU10040
Encompass_Preferred
Mouse Growth Hormone (GH) CLIA Kit for detecting GH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 1.91pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06880. Gene ID: 14599. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5817