Learn More
Biomatik Corporation Mouse Growth Hormone Receptor (GHR) ELISA Kit
Supplier:  Biomatik Corporation EKU04615
Encompass_Preferred
Mouse Growth Hormone Receptor (GHR) ELISA Kit for detecting GHR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16882. Gene ID: 14600. Target Synonyms: GHBP; Somatotropin receptor; Growth hormone-binding protein; Serum-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8943