Learn More
Crystal Chem Inc Mouse Glycated Serum Proteins (GSP) Assay Kit, 96-tests
SDP
Catalog No. 501947704
Encompass_Preferred
The Mouse Glycated Serum Protein Assay is used to quantify levels of glycated serum protein in mouse serum in less than 10 min. Highlights: Kit uses sample size of only 20µL, ideal for mouse applications. Complementary to HbA1c in screening of diabetes. Excellent correlation to ELISA method. Designed for mouse samples. Less than 15 minute procedure. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7704 Supplier Crystal Chem Inc Supplier No. 80420
The Mouse Glycated Serum Protein Assay is used to quantify levels of glycated serum protein in mouse serum in less than 10 min. Highlights: Kit uses sample size of only 20µL, ideal for mouse applications. Complementary to HbA1c in screening of diabetes. Excellent correlation to ELISA method. Designed for mouse samples. Less than 15 minute procedure. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.