Learn More
Biomatik Corporation Mouse Guanylate Binding Protein 2, Interferon Inducible (GBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU12229
Encompass_Preferred
Mouse Guanylate Binding Protein 2, Interferon Inducible (GBP2) ELISA Kit for detecting GBP2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0E6. Gene ID: 14469. Target Synonyms: GTP-binding protein 2; Guanine nucleotide-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4836