Learn More
BIOHIPPO Mouse GZMB Microsample
SDP
Catalog No. 5000315238
Encompass
Mouse GZMB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 81 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GZMB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GZMB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GZMB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15238 Supplier BIOHIPPO Supplier No. BHE15212217
Mouse GZMB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 81 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse GZMB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse GZMB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse GZMB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color