Learn More
ACROBiosystems PE-Labeled Mouse H-2Kb&B2M Tetramer Protein (Peptide free)
Catalog No. 502719521
Encompass_Preferred
PE-Labeled Mouse H-2Kb&B2M Tetramer Protein (H2M-MP2H7) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 305 (H-2Kb) & Ile 21 - Met 119 (B2M) (Accession # P01901 (H-2Kb) & P01887 (B2M)).
Catalog No. 50-271-9521 Supplier ACROBiosystems Supplier No. H2MMP2H725UG
PE-Labeled Mouse H-2Kb&B2M Tetramer Protein (H2M-MP2H7) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 305 (H-2Kb) & Ile 21 - Met 119 (B2M) (Accession # P01901 (H-2Kb) & P01887 (B2M)).