Learn More
ACROBiosystems Biotinylated Mouse H-2Kb&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719526
Encompass_Preferred
Biotinylated Mouse H-2Kb&B2M Monomer Protein (H2M-M82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 305 (H-2Kb) & Ile 21 - Met 119 (B2M) (Accession # P01901 (H-2Kb) & P01887 (B2M)).
Catalog No. 50-271-9526 Supplier ACROBiosystems Supplier No. H2MM82E7200UG
Biotinylated Mouse H-2Kb&B2M Monomer Protein (H2M-M82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 305 (H-2Kb) & Ile 21 - Met 119 (B2M) (Accession # P01901 (H-2Kb) & P01887 (B2M)).