Learn More
World Precision Instrument Replacement Tips for Rodent Handling Forceps (503370)
SDP
Supplier:  World Precision Instrument 504645
Encompass
Rubber-tipped forceps for handling rodents are designed for delicate and precise handling of small animals like mice and rats. These forceps feature a unique design with rubber tips to ensure a secure yet gentle grip, reducing the risk of injury or discomfort to the animals. Rubber-tipped forceps are made from surgical grade stainless steel, which provides durability and strength. The ringed tips of these forceps have replaceable inserts made of soft, non-slip rubber. The rubber grippers ensure a secure grasp without causing damage to the skin or fur of the animals. The forceps are 21 cm (8.25”) long and are straight. They are designed to provide precise control over the animal without causing unnecessary stress or harm. The rubber tips offer a non-slip grip, preventing the animal from slipping or being injured during handling. This feature is particularly useful when holding rodents for various procedures, such as injections, examinations, or minor surgeries.
Catalog No. NC1565793