Learn More
Fine Science Tools Mouse Handling Forceps Replacement Tips, 3 cm, Pair
SDP
Catalog No. NC1288950
Encompass
Mouse Handling Forceps Replacement Tips 1 Pair Tip Shape: Straight Tips: Serrated Tip Dimensions: 15 x 20mm Length: 3cm
Catalog No. NC1288950 Supplier Fine Science Tools Supplier No. 1103690
Mouse Handling Forceps Replacement Tips 1 Pair Tip Shape: Straight Tips: Serrated Tip Dimensions: 15 x 20mm Length: 3cm