Learn More
Sino Biological Recombinant Anti-TIM-1/HAVCR1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50321R15220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TIM-1 / KIM-1 / HACVR (rM TIM-1 / KIM-1 / HACVR; Catalog#50321-M07H; AAL35774.1; Tyr22-Thr212).
Catalog No. 50-240-9848