Learn More
Biomatik Corporation Mouse Heat Shock 10kDa Protein 1 (HSP10) ELISA Kit
Supplier:  Biomatik Corporation EKU04641
Encompass_Preferred
Mouse Heat Shock 10kDa Protein 1 (HSP10) ELISA Kit for detecting HSP10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: q64433. Gene ID: 15528. Target Synonyms: HSPE1; CPN10; EPF; GROES; Chaperonin 10; Early Pregnancy Factor; 10 kDa heat shock protein, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5925