Learn More
Biomatik Corporation Mouse Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit
Supplier:  Biomatik Corporation EKU04655
Encompass_Preferred
Mouse Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit for detecting HSPA1A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61696. Gene ID: 193740. Target Synonyms: HSP70-1; HSP-A1A; HSPA1; HSP70-2; HSX70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6314