Learn More
Biomatik Corporation Mouse Heat Shock Protein 60 (Hsp60) CLIA Kit
Supplier:  Biomatik Corporation EKU11932
Encompass_Preferred
Mouse Heat Shock Protein 60 (Hsp60) CLIA Kit for detecting Hsp60 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.04ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P63038. Gene ID: 15510. Target Synonyms: HSPD1; GROEL; CPN60; HuCHA60; Heat Shock 60kD Protein 1, Chaperonin; Spastic Paraplegia 13,Autosomal Dominant; 60 kDa chaperonin; Mitochondrial matrix protein P1; P60 lymphocyte protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8062