Learn More
Biomatik Corporation Mouse Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit
Supplier:  Biomatik Corporation EKU04682
Encompass_Preferred
Mouse Heat Shock Protein 90kDa Beta 1 (HSP90b1) ELISA Kit for detecting HSP90b1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08113. Gene ID: 22027. Target Synonyms: HSP90-B1; ECGP; GP96; GRP94; TRA1; Glycoprotein 96; Tumor Rejection Antigen (gp96) 1;94 kDa glucose-regulated protein; Endoplasmin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7388