Learn More
ACROBiosystems Mouse Hemagglutinin HA 100ug
Catalog No. 502534891
Encompass_Preferred
Influenza commonly known as the flu is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae the influenza viruses. The virus is divided into three main types (Influenzavirus A Influenzavirus B and Influenzavirus C) which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA) which are the most important targets for the immune system. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle which makes it a great target for vaccine studies.
Catalog No. 50-253-4891 Supplier ACROBiosystems Supplier No. HA1Y159100UG
Influenza commonly known as the flu is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae the influenza viruses. The virus is divided into three main types (Influenzavirus A Influenzavirus B and Influenzavirus C) which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA) which are the most important targets for the immune system. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle which makes it a great target for vaccine studies.