Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Hematopoetic Stem Cell SLAM Code Panel, BD

Catalog No. BDB562461 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562461 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562461 Supplier BD Biosciences Supplier No. 562461

missing translation for 'validMainframeMsg'

The TC15-12F12.2 monoclonal antibody specifically binds to mouse CD150, also known as SLAM (signaling lymphocyte activation molecule). CD150 is a type 1 transmembrane glycoprotein that is a member of the CD2 subfamily of the Ig superfamily. It is encoded by the Slamf1 (signaling lymphocytic activation molecule family member 1) gene. CD150 is differentially expressed on subsets of thymocytes, T and B lymphocytes, dendritic cells, macrophages, and endothelial cells. SLAM plays multiple roles in innate and adaptive immunity serving as an adhesion molecule and/or coreceptor. CD150-mediated costimulation of TCR-activated T cells reportedly results in the increased production of IFN-γ by Th1 cells and is required for IL-4 production by T follicular helper cells. CD150 also plays important roles in hematopoietic cell developmental pathways. CD150 is differentially expressed by self-renewing adult hematopoietic stem cells (HSC) that are CD150+ whereas non-multipotent hematopoietic progenitor cells are CD150-. Utilizing additional cell surface markers, lineage-negative CD150+CD48-CD41- cell fractions are reported to be highly enriched for adult HSC.

Host Species: Rat, Hamster, Rat
Clone: TC15-12F12.2, HM48-1, MWReg30
Isotype: IgG2a, λ, IgG1, λ 3, IgG1, κ
Species Reactivity: Ms

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC/PE/Alexa Fluor 647
Host Species Rat, Hamster
Quantity 50 μg
Isotype IgG2a λ, IgG1 λ3, IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.