Learn More
American Research Products Inc Mouse heme oXygenase 1 HO 1 E
Catalog No. 5000329689
Encompass
Mouse heme oXygenase 1 HO-1 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29689 Supplier American Research Products Inc Supplier No. CSBE08268M
Mouse heme oXygenase 1 HO-1 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich