Learn More
Biomatik Corporation Mouse Hemicentin 1 (HMCN1) ELISA Kit
Supplier:  Biomatik Corporation EKU08455
Encompass_Preferred
Mouse Hemicentin 1 (HMCN1) ELISA Kit for detecting HMCN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C716. Target Synonyms: ARMD1; AXPC1; FBLN6; FIBL6; Fibulin 6; Age Related Macular Degeneration 1 (Senile Macular Degeneration). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8664