Learn More
Cusabio Technology LLC MOUSE HEMOGLOBIN 1G
Catalog No. NC2186698
Encompass_Preferred
Mouse Hemoglobin is a native protein with full length. This protein is purified from mouse erythrocytes. Its purity was determined by SDS-PAGE and reached up to 95%. It is presented the mass molecule around 14.4 kDa visualized on the gel. It has been validated in ELISA, WB and SDS-PAGE applications.Hemoglobin is the protein molecule in red blood cells involved in oxygen transport from the lung to the various peripheral tissues and returns carbon dioxide from the tissues back to the lungs. It is made up of four chains. Each of these chains contains a compound called heme, which in turn contains iron, which is what transports oxygen in the bloodstream. A healthy individual has 12 to 20 grams of hemoglobin in every 100 ml of blood.
Catalog No. NC2186698 Supplier Cusabio Technology LLC Supplier No. CSBNP004901M
Mouse Hemoglobin is a native protein with full length. This protein is purified from mouse erythrocytes. Its purity was determined by SDS-PAGE and reached up to 95%. It is presented the mass molecule around 14.4 kDa visualized on the gel. It has been validated in ELISA, WB and SDS-PAGE applications.Hemoglobin is the protein molecule in red blood cells involved in oxygen transport from the lung to the various peripheral tissues and returns carbon dioxide from the tissues back to the lungs. It is made up of four chains. Each of these chains contains a compound called heme, which in turn contains iron, which is what transports oxygen in the bloodstream. A healthy individual has 12 to 20 grams of hemoglobin in every 100 ml of blood.