Learn More
Abcam Mouse Hemoglobin ELISA Kit (Hba), 1 x 96 Tests
Supplier:  Abcam AB2545171X96TESTS
Encompass_Preferred
Mouse Hemoglobin ELISA Kit (Hba) is a single-wash 90-min Simplestep used to quantify Mouse Hemoglobin (Hba) with a sensitivity of 0.1 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2076237