Learn More
Biomatik Corporation Mouse Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) CLIA Kit
Supplier:  Biomatik Corporation EKU09663
Encompass_Preferred
Mouse Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) CLIA Kit for detecting HBEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q06186. Gene ID: 15200. Target Synonyms: HB-EGF; DTR; DTS; DTSF; HEGFL; Diphtheria Toxin Receptor; Proheparin-binding EGF-like growth factor; Diphtheria toxin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6652