Learn More
Biomatik Corporation Mouse Heparin Cofactor II (HCII) ELISA Kit
Supplier:  Biomatik Corporation EKU04744
Encompass_Preferred
Mouse Heparin Cofactor II (HCII) ELISA Kit for detecting HCII in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49182. Gene ID: 15160. Target Synonyms: SERPIND1; HC2; HCF2; HC II; HLS2; LS2; Serpin D1; Serpin Peptidase Inhibitor Clade D Member 1; Protease inhibitor leuserpin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5759