Learn More
ATCC Mouse; hepatitis Virus
Catalog No. 502384017
Encompass
Murine hepatitis virus strain MHV 1 (Parkes) can be propagated in NCTC clone 1469 [derivative of NCTC 721] cells (ATCC CCL-9.1) or neonatal mice. This strain was isolated in 1950 from liver and kidney tissues from ill mice in London, England.
Catalog No. 50-238-4017 Supplier ATCC Supplier No. VR261
Murine hepatitis virus strain MHV 1 (Parkes) can be propagated in NCTC clone 1469 [derivative of NCTC 721] cells (ATCC CCL-9.1) or neonatal mice. This strain was isolated in 1950 from liver and kidney tissues from ill mice in London, England.