Learn More
American Research Products Inc Mouse hepatocyte growth factor
Catalog No. 5000326799
Encompass
Mouse hepatocyte growth factor HGF ELISA kit from CUSABIO Detection Range 125 pg/ml-8000 pg/ml Sensitivity 31 25 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26799 Supplier American Research Products Inc Supplier No. CSBE07347M
Mouse hepatocyte growth factor HGF ELISA kit from CUSABIO Detection Range 125 pg/ml-8000 pg/ml Sensitivity 31 25 pg/ml Sample serum plasma tissue homogenates Method Sandwich