Learn More
American Research Products Inc Mouse Hepatocyte Growth Factor
Catalog No. 5000327466
Encompass
Mouse Hepatocyte Growth Factor Receptor (C-MET/HGFR) ELISA Kit from CUSABIO Detection Range 0 078 ng/mL-5 ng/mL Sensitivity 0 02 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27466 Supplier American Research Products Inc Supplier No. CSBE13492M
Add to Cart
Add to Cart
Mouse Hepatocyte Growth Factor Receptor (C-MET/HGFR) ELISA Kit from CUSABIO Detection Range 0 078 ng/mL-5 ng/mL Sensitivity 0 02 ng/mL Sample serum plasma tissue homogenates Method Sandwich