Learn More
American Research Products Inc Mouse Hepatocyte growth factor
Catalog No. 5000327063
Encompass
Mouse Hepatocyte growth factor actiVator (HGFAC) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27063 Supplier American Research Products Inc Supplier No. CSBEL010328MO
Mouse Hepatocyte growth factor actiVator (HGFAC) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich