Learn More
ACROBiosystems Mouse Her2 480tests
Catalog No. 502535113
Encompass_Preferred
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2 HER-2HER-2 /neu NEU NGLTKR1 and c-erb B2and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development cancer communication at the neuromuscular junction and regulation of cell growth and differentiation .
Catalog No. 50-253-5113 Supplier ACROBiosystems Supplier No. EHMV1480TESTS
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2 HER-2HER-2 /neu NEU NGLTKR1 and c-erb B2and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development cancer communication at the neuromuscular junction and regulation of cell growth and differentiation .