Learn More
Biomatik Corporation Mouse Hexokinase 2 (HK2) ELISA Kit
Supplier:  Biomatik Corporation EKU09160
Encompass_Preferred
Mouse Hexokinase 2 (HK2) ELISA Kit for detecting HK2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08528. Gene ID: 15277. Target Synonyms: HK II; Muscle form hexokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6869