Learn More
Raybiotech Inc Mouse HGF ELISA Kit
Supplier:  Raybiotech Inc ELMHGFCL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 400 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse HGF.
Catalog No. 50-195-0007