Learn More
Gibco™ Mouse HGF Recombinant Protein, PeproTech®
Description
315-23-1MG will be provided as 4 x 250 μg (315-23-250UG). 315-23-500UG will be provided as 2 x 250 μg (315-23-250UG). Recombinant Murine HGF is a 79.3 kDa polypeptide consisting of 696 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | Q08048 |
| For Use With (Application) | Bioactivity, Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 15234 |
| Molecular Weight (g/mol) | 79.3 kDa |
| Name | Mouse HGF |
| Purification Method | Purified |
| Quantity | 5 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.