Learn More
American Research Products Inc Mouse High density lipoprotein
Supplier:  American Research Products Inc CSBE12840M
Encompass
Mouse High density lipoprotein HDL ELISA Kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 8 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29989