Learn More
American Research Products Inc Mouse High density lipoprotein
Catalog No. 5000329800
Encompass
Mouse High density lipoprotein cholesterol HDL-C ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29800 Supplier American Research Products Inc Supplier No. CSBE12874M
Mouse High density lipoprotein cholesterol HDL-C ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich