Learn More
American Research Products Inc Mouse High mobility group prot
Catalog No. 5000329691
Encompass
Mouse High mobility group protein B1 HMGB-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29691 Supplier American Research Products Inc Supplier No. CSBE08225M
Mouse High mobility group protein B1 HMGB-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich