Learn More
Biomatik Corporation Mouse High Molecular Weight Kininogen (HMWK) ELISA Kit
Supplier:  Biomatik Corporation EKU10906
Encompass_Preferred
Mouse High Molecular Weight Kininogen (HMWK) ELISA Kit for detecting HMWK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.288ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08677. Gene ID: 16644. Target Synonyms: Williams-Fitzgerald-Flaujeac Factor; Fitzgerald Factor; HMWK-Kallikrein Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7172