Learn More
Alpco Ltd Mouse High Range Insulin ELISA, Superior Sensitivity
SDP
Supplier:  Alpco Ltd 80INSMSHE01
Encompass_Preferred
The Mouse High Range Insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes from as little as 5 µL of serum or plasma sample.
  • Low sample volume of 5 µL
  • Broad dynamic range of 3-150 ng/mL
  • Superior sensitivity of 1.99 ng/mL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC1075404