Learn More
ACROBiosystems Biotinylated Anti-His Tag Antibody(AY63), Fab
Catalog No. 502329148
Encompass_Preferred
Biotinylated Anti-His Tag Antibody(AY63), Fab is a Fab fragment of a mouse monoclonal antibody recombinantly expressed from human 293 cells (HEK293). The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with a poly-his tagged protein.
Catalog No. 50-232-9148 Supplier ACROBiosystems Supplier No. HISBVM539100UG
Biotinylated Anti-His Tag Antibody(AY63), Fab is a Fab fragment of a mouse monoclonal antibody recombinantly expressed from human 293 cells (HEK293). The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with a poly-his tagged protein.