Learn More
ACROBiosystems Mouse His 1mg
Catalog No. 502534790
Encompass_Preferred
His tag is a very small molecular weight tag usually composed of 6-10 histidine (His). It is one of the commonly used tags for protein purification and detection. Due to its small molecular weight fusion into the target protein has almost no effect on the structure and characteristics of the protein. Anti-his tag antibody can accurately detect locate and purify His tag fusion protein so as to provide convenience for the vast number of researchers.
Catalog No. 50-253-4790 Supplier ACROBiosystems Supplier No. HISFM5351MG
His tag is a very small molecular weight tag usually composed of 6-10 histidine (His). It is one of the commonly used tags for protein purification and detection. Due to its small molecular weight fusion into the target protein has almost no effect on the structure and characteristics of the protein. Anti-his tag antibody can accurately detect locate and purify His tag fusion protein so as to provide convenience for the vast number of researchers.