Learn More
Biomatik Corporation Mouse Histamine Receptor H4 (HRH4) ELISA Kit
Supplier:  Biomatik Corporation EKU12292
Encompass_Preferred
Mouse Histamine Receptor H4 (HRH4) ELISA Kit for detecting HRH4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 246.9-20,000pg/ml. Sensitivity: 92.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q91ZY2. Gene ID: 225192. Target Synonyms: GPCR105; GPRv53; H4R; HH4R; H4; AXOR35; BG26; G-protein coupled receptor 105. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7990